Acyclovir Related Compound G
Synonym(s):
9-[(2-Acetoxyethoxy)methyl]-N2-acetylguanine, 2-{[2-(Acetylamino)-6-oxo-1,6-dihydro-9H-purin-9-yl]methoxy}ethyl acetate
Empirical Formula (Hill Notation):
C12H15N5O5
CAS Number:
Molecular Weight:
309.28
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2