
Acriflavine hydrochloride

Synonym(s):
3,6-Diamino-10-methylacridinium chloride hydrochloride, Euflavine
CAS Number:
Products
Product Category
Brand
Color
Melting Point (°C)
Physical Form
Chemical Composition
Quality Segment
Application
Technique
Showing 1-1 of 1