Skip to Content
Merck
α,β-Thujone phyproof® Reference Substance

α,β-Thujone

α,β-Thujone phyproof® Reference Substance
Synonym(s):
(−)-1-Isopropyl-4-methylbicyclo[3.1.0]hexan-3-one
Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Chemical Composition
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2