![6-Carboxyfluorescein diacetate ≥95% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/358/176/fd895d92-9090-4641-8e29-41f249ab8e97/640/fd895d92-9090-4641-8e29-41f249ab8e97.png)
6-Carboxyfluorescein diacetate
![6-Carboxyfluorescein diacetate ≥95% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/358/176/fd895d92-9090-4641-8e29-41f249ab8e97/640/fd895d92-9090-4641-8e29-41f249ab8e97.png)
Synonym(s):
6-CFDA, 6-Carboxy-di-O-acetylfluorescein
Empirical Formula (Hill Notation):
C25H16O9
CAS Number:
Molecular Weight:
460.39
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing