5-Methyl-1,3-cyclohexanedione
Linear Formula:
CH3C6H7(=O)2
CAS Number:
Molecular Weight:
126.15
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing