![5-Bromo-4-chloro-3-indolyl β-D-glucopyranoside ≥97%](/deepweb/assets/sigmaaldrich/product/structures/221/547/d16f7163-7205-40f9-9290-6f62a22e383b/640/d16f7163-7205-40f9-9290-6f62a22e383b.png)
5-Bromo-4-chloro-3-indolyl β-D-glucopyranoside
![5-Bromo-4-chloro-3-indolyl β-D-glucopyranoside ≥97%](/deepweb/assets/sigmaaldrich/product/structures/221/547/d16f7163-7205-40f9-9290-6f62a22e383b/640/d16f7163-7205-40f9-9290-6f62a22e383b.png)
Synonym(s):
X-Glc, X-glucoside
Empirical Formula (Hill Notation):
C14H15BrClNO6
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing