Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
5-Aminosalicylic acid
Synonym(s):
5-AS, 5-Amino-2-hydroxybenzoic acid, Mesalamine
Linear Formula:
H
2
NC
6
H
3
-2-(OH)CO
2
H
CAS Number:
89-57-6
Molecular Weight:
153.14
EC Number:
201-919-1
Products
Product Category
amino acids (1)
substrates (1)
Brand
Sigma-Aldrich (2)
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
A3537
5-Aminosalicylic acid,
powder, ≥99%
Expand
View Pricing
A79809
5-Aminosalicylic acid,
95%
Expand
View Pricing