5(6)-Carboxyfluorescein N-hydroxysuccinimide ester
Synonym(s):
5(6)-FAM SE, FLUOS
Empirical Formula (Hill Notation):
C25H15NO9
CAS Number:
Molecular Weight:
473.39
Products
Brand
Formula Weight
Physical Form
Product Line
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing