Skip to Content
Merck
5,10,15,20-Tetraphenyl-21H,23H-porphine zinc

5,10,15,20-Tetraphenyl-21H,23H-porphine zinc

5,10,15,20-Tetraphenyl-21H,23H-porphine zinc
Synonym(s):
meso-Tetraphenylporphine, TPP, Zinc TPP
Empirical Formula (Hill Notation):
C44H28N4Zn
CAS Number:
Molecular Weight:
678.11

Products

Brand
Formula Weight
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing