![5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphine cobalt(II)](/deepweb/assets/sigmaaldrich/product/structures/601/003/f57b2dfd-0b73-4ab2-bc8a-a18aa80c4121/640/f57b2dfd-0b73-4ab2-bc8a-a18aa80c4121.png)
5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphine cobalt(II)
![5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphine cobalt(II)](/deepweb/assets/sigmaaldrich/product/structures/601/003/f57b2dfd-0b73-4ab2-bc8a-a18aa80c4121/640/f57b2dfd-0b73-4ab2-bc8a-a18aa80c4121.png)
Empirical Formula (Hill Notation):
C48H36CoN4O4
CAS Number:
Molecular Weight:
791.76
Products
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing