![4-tert-Octylphenol-ring-13C6 solution 10 μg/mL in acetone, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/119/380/34812c3b-28d5-469f-bff4-5bd16daa4b6c/640/34812c3b-28d5-469f-bff4-5bd16daa4b6c.png)
4-tert-Octylphenol-ring-13C6 solution
![4-tert-Octylphenol-ring-13C6 solution 10 μg/mL in acetone, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/119/380/34812c3b-28d5-469f-bff4-5bd16daa4b6c/640/34812c3b-28d5-469f-bff4-5bd16daa4b6c.png)
Synonym(s):
4-(1,1,3,3-Tetramethylbutyl)phenol-13C6, 4-tert-OP-13C6
Empirical Formula (Hill Notation):
13C6C8H22O
Products
Brand
Formula Weight
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2