Skip to Content
Products
Cart
0
AR
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
4-Nitrophenyl α-L-fucopyranoside
Empirical Formula (Hill Notation):
C
12
H
15
NO
7
CAS Number:
10231-84-2
Molecular Weight:
285.25
EC Number:
233-553-3
Products
Product Category
substrates (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Physical Form
Purity
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
N3628
4-Nitrophenyl α-L-fucopyranoside,
≥98% (TLC), powder
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
Español - ES
English - EN
Learn More