Skip to Content
Merck
4-Nitrophenol PESTANAL®, analytical standard

4-Nitrophenol

4-Nitrophenol PESTANAL®, analytical standard
Synonym(s):
p-Nitrophenol, p-Nitrophenol solution, 4-Nitrophenol
Linear Formula:
O2NC6H4OH
CAS Number:
Molecular Weight:
139.11
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Feature
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing