
4-Nitro-L-phenylalanine monohydrate

Linear Formula:
O2NC6H4CH2CH(NH2)CO2H · H2O
CAS Number:
Molecular Weight:
228.20
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1