![4-Chloro-7-sulfobenzofurazan ammonium salt](/deepweb/assets/sigmaaldrich/product/structures/295/987/66a7016b-f853-4f5a-9dd0-2778bf36c82e/640/66a7016b-f853-4f5a-9dd0-2778bf36c82e.png)
4-Chloro-7-sulfobenzofurazan ammonium salt
![4-Chloro-7-sulfobenzofurazan ammonium salt](/deepweb/assets/sigmaaldrich/product/structures/295/987/66a7016b-f853-4f5a-9dd0-2778bf36c82e/640/66a7016b-f853-4f5a-9dd0-2778bf36c82e.png)
Synonym(s):
4-Chloro-7-sulfobenz-2-oxa-1,3-diazole ammonium salt, 7-Chloro-2,1,3-benzoxadiazole-4-sulfonic acid ammonium salt, 7-Chloro-4-benzofurazansulfonic acid ammonium salt, SBF-Chloride
Linear Formula:
C6H3ClN2O4S · NH3
CAS Number:
Molecular Weight:
251.65
Products
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing