Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
4-Chloro-2-isopropyl-5-methylphenol
Synonym(s):
Chlorothymol
Linear Formula:
(CH
3
)
2
CHC
6
H
2
(Cl)(CH
3
)OH
CAS Number:
89-68-9
Molecular Weight:
184.66
EC Number:
201-930-1
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
261416
4-Chloro-2-isopropyl-5-methylphenol,
99%
Expand
View Pricing