Skip to Content
Merck
4-Carboxy-TEMPO, free radical 97%

4-Carboxy-TEMPO, free radical

4-Carboxy-TEMPO, free radical 97%
Synonym(s):
4-Carboxy-2,2,6,6-tetramethylpiperidine 1-oxyl, 4-Carboxy-2,2,6,6-tetramethylpiperidinyloxy, free radical, 4-Carboxy-TEMPO
Empirical Formula (Hill Notation):
C10H18NO3
CAS Number:
Molecular Weight:
200.25

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing