4-Bromomethyl-7-methoxycoumarin
Synonym(s):
BMC, Br-Mmc
Empirical Formula (Hill Notation):
C11H9BrO3
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing