4-Amino-TEMPO, free radical
Synonym(s):
4-Amino-2,2,6,6-tetramethylpiperidine-1-oxyl, 4-Amino-2,2,6,6-tetramethylpiperidinyloxy, free radical, 4-Amino-TEMPO
Empirical Formula (Hill Notation):
C9H19N2O
Products
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing