Skip to Content
Merck
4-Amino-5-phenyl-4H-1,2,4-triazole-3-thiol 97%

4-Amino-5-phenyl-4H-1,2,4-triazole-3-thiol

4-Amino-5-phenyl-4H-1,2,4-triazole-3-thiol 97%
Empirical Formula (Hill Notation):
C8H8N4S
CAS Number:
Molecular Weight:
192.24

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing