Skip to Content
Merck
4-Acetamido-4′-isothiocyanato-2,2′-stilbenedisulfonic acid disodium salt hydrate ≥80%

4-Acetamido-4′-isothiocyanato-2,2′-stilbenedisulfonic acid disodium salt hydrate

4-Acetamido-4′-isothiocyanato-2,2′-stilbenedisulfonic acid disodium salt hydrate ≥80%
Synonym(s):
Disodium 4-acetamido-4′-isothiocyanato-stilben-2,2′-disulfonate, SITS
Empirical Formula (Hill Notation):
C17H12N2Na2O7S3 · xH2O
CAS Number:
Molecular Weight:
498.46 (anhydrous basis)
EC Number:

Products

Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1