Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
4,5-Dicyanoimidazole
Synonym(s):
4,5-Imidazoledicarbonitrile
Empirical Formula (Hill Notation):
C
5
H
2
N
4
CAS Number:
1122-28-7
Molecular Weight:
118.10
EC Number:
214-344-6
Products
Product Category
building blocks (2)
Brand
Sigma-Aldrich (2)
Color
Formula Weight
Melting Point (°C)
Product Line
Purity
Functional Group
Markush Class
Markush Group
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
554030
4,5-Dicyanoimidazole,
99.0%
Expand
View Pricing
324132
4,5-Dicyanoimidazole,
95%
Expand
View Pricing
M070000
4,5-Dicyanoimidazole,
Expand
View Pricing