Skip to Content
Merck
4,4′-Bis(carbazol-9-yl)biphenyl ≥99% (HPLC)

4,4′-Bis(carbazol-9-yl)biphenyl

4,4′-Bis(carbazol-9-yl)biphenyl ≥99% (HPLC)
Synonym(s):
4,4′-Bis(N-carbazolyl)-1,1′-biphenyl, 4,4′-Bis(9-carbazolyl)-1,1′-biphenyl, 4,4-N,N′-Dicarbazole-1,1′-biphenyl, CBP, DCBP
Empirical Formula (Hill Notation):
C36H24N2
CAS Number:
Molecular Weight:
484.59

Products

Brand
Formula Weight
Melting Point (°C)
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing