Skip to Content
Merck
4-[2-(5-Chloro-2-methoxybenzamido)ethyl]benzenesulfonamide British Pharmacopoeia (BP) Reference Standard

4-[2-(5-Chloro-2-methoxybenzamido)ethyl]benzenesulfonamide

4-[2-(5-Chloro-2-methoxybenzamido)ethyl]benzenesulfonamide British Pharmacopoeia (BP) Reference Standard
Synonym(s):
5-Chloro-2-methoxy-N-[2-(4-sulfamoylphenyl)ethyl]benzamide, 4-[2-(5-Chloro-2-methoxybenzamido)ethyl]phenylsulfonamide
Linear Formula:
ClC6H3(OCH3)CONHCH2C6H4SO2NH2
CAS Number:
Molecular Weight:
368.84

Products

Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade