4-(1HTetrazol-5-yl)phenylboronic acid
Empirical Formula (Hill Notation):
C7H7BN4O2
CAS Number:
Molecular Weight:
189.97
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing