
3-Phenylbenzyl bromide

Synonym(s):
3-Bromomethylbiphenyl
Empirical Formula (Hill Notation):
C13H11Br
CAS Number:
Molecular Weight:
247.13
Products
Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing