![3-(Decyldimethylammonio)propanesulfonate inner salt zwitterionic detergent](/deepweb/assets/sigmaaldrich/product/structures/389/627/1350b348-414a-40fa-a57e-1068ce4c84a7/640/1350b348-414a-40fa-a57e-1068ce4c84a7.png)
3-(Decyldimethylammonio)propanesulfonate inner salt
![3-(Decyldimethylammonio)propanesulfonate inner salt zwitterionic detergent](/deepweb/assets/sigmaaldrich/product/structures/389/627/1350b348-414a-40fa-a57e-1068ce4c84a7/640/1350b348-414a-40fa-a57e-1068ce4c84a7.png)
Synonym(s):
Caprylyl sulfobetaine, SB3-10
Linear Formula:
CH3(CH2)9N+(CH3)2CH2CH2CH2SO3-
CAS Number:
Molecular Weight:
307.49
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing