Skip to Content
Merck
3-Bromo-1-(trimethylsilyl)-1-propyne 98%

3-Bromo-1-(trimethylsilyl)-1-propyne

3-Bromo-1-(trimethylsilyl)-1-propyne 98%
Synonym(s):
3-(Trimethylsilyl)propargyl bromide, 3-Bromo-1-(trimethylsilyl)-1-propyne
Linear Formula:
BrCH2C≡CSi(CH3)3
CAS Number:
Molecular Weight:
191.14

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing