Skip to Content
Merck
3-Aminophenylboronic acid hemisulfate salt ≥95%

3-Aminophenylboronic acid hemisulfate salt

3-Aminophenylboronic acid hemisulfate salt ≥95%
Synonym(s):
(3-Aminophenyl)boric acid hemisulfate, (3-aminophenyl)-boronic acid sulfate (2:1), (m-Aminophenyl)boronic acid hemisulfate, 3-Aminobenzeneboronic acid hemisulfate salt, Bis[3-dihydroxyboranyl)benzenaminium] sulfate
Linear Formula:
H2NC6H4B(OH)2 · 0.5H2SO4
CAS Number:
Molecular Weight:
185.98
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing