Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
3-aminomethylphenylboronic acid, pinacol ester hydrochloride
Empirical Formula (Hill Notation):
C
13
H
21
BClNO
2
CAS Number:
380430-65-9
Molecular Weight:
269.58
Products
Brand
Sigma-Aldrich (3)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
741205
3-aminomethylphenylboronic acid, pinacol ester hydrochloride,
97%
Expand
View Pricing
CDS005517
3-aminomethylphenylboronic acid, pinacol ester hydrochloride,
Aldrich
CPR
Expand
View Pricing
BML00288
3-aminomethylphenylboronic acid, pinacol ester hydrochloride,
Aldrich
CPR
Expand
View Pricing