[3-(4-fluoro-phenyl)-isoxazol-5-yl]-methanol
Empirical Formula (Hill Notation):
C10H8FNO2
Molecular Weight:
193.17
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing