Skip to Content
Merck
Thiamine hydrochloride European Pharmacopoeia (EP) Reference Standard

Thiamine hydrochloride

Thiamine hydrochloride European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Aneurine hydrochloride, Vitamin B1 hydrochloride, Thiamine hydrochloride, Thiamine, Hydrochloride, Thiamine chloride hydrochloride
Empirical Formula (Hill Notation):
C12H17ClN4OS · HCl
CAS Number:
Molecular Weight:
337.27

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique