Skip to Content
Merck
2-(Tetrazol-5-yl)phenylboronic acid ≥95%

2-(Tetrazol-5-yl)phenylboronic acid

2-(Tetrazol-5-yl)phenylboronic acid ≥95%
Empirical Formula (Hill Notation):
C7H7BN4O2
CAS Number:
Molecular Weight:
189.97

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing