Skip to Content
Merck
(2S)-2-(5-Iodo-6-methoxynaphthalen-2-yl)propanoic acid pharmaceutical secondary standard, certified reference material

(2S)-2-(5-Iodo-6-methoxynaphthalen-2-yl)propanoic acid

(2S)-2-(5-Iodo-6-methoxynaphthalen-2-yl)propanoic acid pharmaceutical secondary standard, certified reference material
Synonym(s):
S)-5-Iodo-6-methoxy-α-methyl-2-naphthaleneacetic acid, (2S)-2-(5-Iodo-6-methoxy-2-naphthyl)propionic acid, (S)-5-Iodo naproxen, Naproxen EP Impurity D
Empirical Formula (Hill Notation):
C14H13IO3
CAS Number:
Molecular Weight:
356.16

Products

Brand
Formula Weight
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing