Skip to Content
Merck
2-Phenylphenol PESTANAL®, analytical standard

2-Phenylphenol

2-Phenylphenol PESTANAL®, analytical standard
Synonym(s):
2-Hydroxybiphenyl
Linear Formula:
C6H5C6H4OH
CAS Number:
Molecular Weight:
170.21
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique