Skip to Content
Merck
2-Methylvaleric acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

2-Methylvaleric acid

2-Methylvaleric acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
(±)-2-Methylvaleric acid, 2-Methylpentanoic acid
Linear Formula:
CH3CH2CH2CH(CH3)CO2H
CAS Number:
Molecular Weight:
116.16

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade