![2-Methylresorcinol analytical standard](/deepweb/assets/sigmaaldrich/product/structures/283/057/2833f382-077b-46d1-b935-ed96c4edc548/640/2833f382-077b-46d1-b935-ed96c4edc548.png)
2-Methylresorcinol
![2-Methylresorcinol analytical standard](/deepweb/assets/sigmaaldrich/product/structures/283/057/2833f382-077b-46d1-b935-ed96c4edc548/640/2833f382-077b-46d1-b935-ed96c4edc548.png)
Synonym(s):
2,6-Dihydroxytoluene
Linear Formula:
CH3C6H3(OH)2
CAS Number:
Molecular Weight:
124.14
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing