Skip to Content
Merck
2-Methylheptanoic acid ≥97%

2-Methylheptanoic acid

2-Methylheptanoic acid ≥97%
Synonym(s):
2-Methylheptanoic acid
Linear Formula:
CH3(CH2)4CH(CH3)CO2H
CAS Number:
Molecular Weight:
144.21

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing