Skip to Content
Merck
2-Methyl-4-nitropyridine N-oxide 97%

2-Methyl-4-nitropyridine N-oxide

2-Methyl-4-nitropyridine N-oxide 97%
Synonym(s):
4-Nitro-2-picoline N-oxide
Empirical Formula (Hill Notation):
C6H6N2O3
CAS Number:
Molecular Weight:
154.12

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing