Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2in1 kit universal vial, short thread autosampler vials
Products
Product Category
vials (2)
Brand
Supelco (2)
Diameter (mm)
ID (mm)
OD (mm)
Volume (ml)
Color
Package Size
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
SU860197
2in1 kit universal vial, short thread autosampler vials,
pkg of 100 ea, volume 0.3 mL, blue polypropylene cap (with 6 mm center hole), clear polypropylene (with integrated 0.3 mL insert)
Expand
View Pricing
SU860196
2in1 kit universal vial, short thread autosampler vials,
pkg of 100 ea, volume 1.5 mL, blue polypropylene cap (open-top), red PTFE/white silicone
Expand
View Pricing