![2′-Hydroxyacetophenone ReagentPlus®, 99%](/deepweb/assets/sigmaaldrich/product/structures/356/394/7f753837-f3cb-4ede-b270-5d16275c5d89/640/7f753837-f3cb-4ede-b270-5d16275c5d89.png)
2′-Hydroxyacetophenone
![2′-Hydroxyacetophenone ReagentPlus®, 99%](/deepweb/assets/sigmaaldrich/product/structures/356/394/7f753837-f3cb-4ede-b270-5d16275c5d89/640/7f753837-f3cb-4ede-b270-5d16275c5d89.png)
Synonym(s):
2-Acetylphenol, 2′-Hydroxyacetophenone
Linear Formula:
2-(OH)C6H4COCH3
CAS Number:
Molecular Weight:
136.15
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing