![2H,2H,3H,3H-Perfluoroundecanoic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/366/749/28b88e2f-fbd6-4740-8b94-3a8474b10ba2/640/28b88e2f-fbd6-4740-8b94-3a8474b10ba2.png)
2H,2H,3H,3H-Perfluoroundecanoic acid
![2H,2H,3H,3H-Perfluoroundecanoic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/366/749/28b88e2f-fbd6-4740-8b94-3a8474b10ba2/640/28b88e2f-fbd6-4740-8b94-3a8474b10ba2.png)
Synonym(s):
4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-Heptadecafluoroundecanoic acid, 2H,2H,3H,3H-Perfluoroundecanoic acid
Linear Formula:
CF3(CF2)7CH2CH2COOH
CAS Number:
Molecular Weight:
492.13
Products
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing