Skip to Content
Merck
2-Fluoroadenine-9-β-D-arabinofuranoside DNA synthesis and methylation inhibitor

2-Fluoroadenine-9-β-D-arabinofuranoside

2-Fluoroadenine-9-β-D-arabinofuranoside DNA synthesis and methylation inhibitor
Synonym(s):
9-β-D-Arabinofuranosyl-2-fluoroadenine, F-ara-A, Fludarabine des-phosphate
Empirical Formula (Hill Notation):
C10H12FN5O4
CAS Number:
Molecular Weight:
285.23
EC Number:

Products

Brand
Formula Weight
Gene Alias
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing