
2-Ethylpentanoic acid

Empirical Formula (Hill Notation):
C7H14O2
CAS Number:
Molecular Weight:
130.18
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing