Skip to Content
Merck
2-Ethylfuran analytical standard

2-Ethylfuran

2-Ethylfuran analytical standard
Empirical Formula (Hill Notation):
C6H8O
CAS Number:
Molecular Weight:
96.13
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing