Skip to Content
Merck
2-Ethyl-4-methylthiazole ≥98%, FG

2-Ethyl-4-methylthiazole

2-Ethyl-4-methylthiazole ≥98%, FG
Empirical Formula (Hill Notation):
C6H9NS
CAS Number:
Molecular Weight:
127.21
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing