Skip to Content
Merck
2-Deoxy-D-galactose 98%

2-Deoxy-D-galactose

2-Deoxy-D-galactose 98%
Synonym(s):
2-Deoxy-D-lyxohexose
Empirical Formula (Hill Notation):
C6H12O5
CAS Number:
Molecular Weight:
164.16
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing