Skip to Content
Merck
2-Chloropyridine-5-acetonitrile 95%

2-Chloropyridine-5-acetonitrile

2-Chloropyridine-5-acetonitrile 95%
Synonym(s):
6-Chloro-3-pyridylacetonitrile, 2-Chloro-5-(cyanomethyl)pyridine
Empirical Formula (Hill Notation):
C7H5ClN2
CAS Number:
Molecular Weight:
152.58

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing