![2-Chlorobutane ≥99%](/deepweb/assets/sigmaaldrich/product/structures/690/967/7808e09f-9bca-4424-a7dc-9205a0701f7c/640/7808e09f-9bca-4424-a7dc-9205a0701f7c.png)
2-Chlorobutane
![2-Chlorobutane ≥99%](/deepweb/assets/sigmaaldrich/product/structures/690/967/7808e09f-9bca-4424-a7dc-9205a0701f7c/640/7808e09f-9bca-4424-a7dc-9205a0701f7c.png)
Synonym(s):
sec-Butyl chloride, 2-Chlorobutane
Linear Formula:
CH3CH2CHClCH3
CAS Number:
Molecular Weight:
92.57
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing