Skip to Content
Merck
2-Bromo-α-ergocryptine methanesulfonate salt solid

2-Bromo-α-ergocryptine methanesulfonate salt

2-Bromo-α-ergocryptine methanesulfonate salt solid
Synonym(s):
(+)-2-Bromo-12′-hydroxy-2′-(1-methylethyl)-5′-(2-methylpropyl)ergotaman-3′,6′-18-trione methanesulfonate salt, (+)-Bromocriptine methanesulfonate salt, Bromocriptine mesylate salt
Empirical Formula (Hill Notation):
C32H40BrN5O5 · CH4SO3
CAS Number:
Molecular Weight:
750.70
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing